Total immersion thermometers
Supplier: Amarell
AMARG10306
Total immersion thermometers
For everyday use, with red spirit filling for extra safety.
  • Enclosed scale made from milk glass
  • Blue reflecting capillary
  • Total immersion
Ø: 7,0 to 8,0 mm
Filling: red spirit