Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

pH Indicator Papers and Test Strips

https://in.vwr.com/stibo/smallweb/std.lang.all/00/41/18990041.jpg

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"11"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  VWR Chemicals
Description:   Dosatest test strips provide precise and rapid pH determination due to the four different colour blocks for each pH-value
Supplier:  VWR Chemicals
Description:   These Dosatest® strips incorporate the colour scale and are ideal for determining pH values for coloured solutions
Supplier:  VWR Chemicals
Description:   Non bleeding special indicator dyes that are covalently bound to the reagent paper and then adhered to plastic strips.
Supplier:  VWR Chemicals
Description:   Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.
UOM:  1 * 50 Tests
Supplier:  VWR Chemicals
Description:   These Duo indicator papers show two different colours for each pH-value.
Supplier:  VWR Chemicals
Description:   Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.
UOM:  1 * 50 Tests
Supplier:  VWR Chemicals
Description:   These rolls (4.8 m × 10 mm) have only 1 colour zone and are useful where high precision of measurement is not necessary.
Catalog Number: (85409.600)
Supplier:  VWR Chemicals
Description:   These tri indicator papers show three different colours for each pH-value.
UOM:  1 * 5 items
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 8  of 11
  1  2  Next