Indicator paper in rolls, DOSATEST®
VWRC70036.601
Indicator paper in rolls, DOSATEST®
DOSATEST® pH indicator test papers give very good precision and are very easy to read because they have 2 colour zones. The hydrophobic barrier between the zones ensures a distinctive separation of colour bands and facilitates the estimation of intermediate values when used with the colour scale provided.
  • Supplied as 5 m × 10 mm rolls (*except the 3 zone 35307.600 which is 6 m × 14 mm)
  • Wide and narrow pH ranges available