Would you like to log in again?
Stoppers for standard ground joints
Supplier: LENZ LABORGLAS GMBH & CO.KG
DURAN®, clear DIN 12252, hollow, hexagonal grip, with drip tip
DURAN®, clear, DIN 12252, solid, from NS 29/32 semi-hollow, octagonal grip
DURAN®, amber, DIN 12252, solid, from NS 29/32 semi-hollow, octagonal grip
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion
  • Autoclavable
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Drip tip
NS Type Pk VWR Catalog Number Quantity
29/32 With drip tip 1 217-9110
Clear glass
NS Type Pk VWR Catalog Number Quantity
10/19 Clear glass 1 217-9122
12/21 Clear glass 1 217-9123
14/23 Clear glass 1 217-9125
19/26 Clear glass 1 217-9126 Unavailable
34/35 Clear glass 1 217-9132
45/40 Clear glass 1 217-9134
85/55 Clear glass 1 217-0772
Amber glass
NS Type Pk VWR Catalog Number Quantity
12/21 Amber glass 1 217-9143
unavailable This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty