Would you like to log in again?
Cleanroom clogs, Medic
Supplier: VWR Collection
TPU (thermoplastic polyurethane)
Ventilation holes in the removable insole. Note: No heel strap fitted.
  • Autoclavable at 134 °C (including insert) or gamma-sterilisable
Colour: White
EN 344 (anti-static)
Log in to see your contract pricing and availability.
  Remember me on this device
Expand All / Collapse All
Pk Size VWR Catalog Number Quantity
1 PAIR 37 (4) 114-2976 Unavailable
unavailable This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty