VWR® Cleanroom Mop Heads
Supplier: VWR
VWR®
129-0074
VWR® Cleanroom Mop Heads
Mops
Made from polyester with PVA cover or NovaLite™. The mop head quickly absorbs the cleaning agents and can be used for large surfaces, such as floors, walls and ceilings. PVA is perfect for the semiconductor and microelectronics industry. This low-lint, quick drying material ensures clean, dry and residue-free surfaces. NovaLite™ has the same properties as PVA, but it can also be used with disinfectants and autoclaved. With an angled head for easy application.
  • Can be used without a bucket or wringer
  • Quick drying, for moisture critical areas
  • Available in a variety of widths, sterile or non sterile
  • Washable, which means it can be reused
Suitable for Class 10, ISO 4, M2.5 and below cleanrooms.
Ordering information: The complete mop consists of the mop head, a matching stainless steel adapter and the mop handle. Please select the required components and order separately.