Would you like to log in again?
PCR sealing mats
Supplier: Brand
Designed to fit Brand PCR plates exactly and reduce evaporation losses by up to 75% compared with conventional systems.
  • Highly flexible
  • Tight sealing
  • Autoclavable
  • Easily pierced by pipette tips
ORDER
Expand All / Collapse All
Description Pk VWR Catalog Number Quantity
96-well mat, TPE 5 211-0003
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty