Would you like to log in again?
Funnels with short stem
Supplier: DURAN Group
Soda glass
  • Good chemical and physical properties
  • Use in applications that do not involve high thermal stresses
  • Quick and easy to clean
With smooth internal walls. Flare angle 60°. Funnel lip is ground and fire-polished.
Certifications: DIN ISO 4798
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Funnel O-Ø Stem O-Ø For filter Ø Stem length Pk VWR Catalog Number Quantity
55 mm 8 mm 70 - 90 mm 55 mm 10 221-0145 Unavailable
unavailable This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty