Would you like to log in again?
Shoe covers, non woven PP
Supplier: VWR Collection
VWR®
Shoe covers made from non woven PP.
  • Laminated
Opening when stretched: 290 mm
L×H: 430×170 mm
*One size fits all
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Colour Size Pk VWR Catalog Number Quantity
Blue U* 300 113-8285
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty