VWR®, Cleaning Brushes for Gas Washing Bottles and Cylinders
Supplier: VWR
VWR®
149-0223EA 884 INR
17252-120 149-0223 149-0202 149-0224
VWR®, Cleaning Brushes for Gas Washing Bottles and Cylinders
Brushes
These cleaning brushes are specially designed for cleaning gas washing bottles, cylinders and large test tubes.
  • Tied tip to reduce the danger of scratching and improve cleaning intensity
  • Wire handle for secure grip
  • Hanging loop