Would you like to log in again?
Bottles, wide neck, with screw cap
Supplier: TECHNICAL TREATMENTS
HDPE, translucent
  • Tough, extra thick walls
  • Screw cap with gasket
  • Good chemical resistance, good temperature resistance
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Capacity Ø ext. Height Neck I-Ø Pk VWR Catalog Number Quantity
300 ml 70 mm 113 mm 48 mm 1 215-2716
700 ml 86 mm 169 mm 48 mm 1 215-2724
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty