Would you like to log in again?
Deep well plates, 384-well, Nunc™ DeepWell™
Supplier: Thermo Scientific
PP, non treated, non sterile, without lid. This deep well plate provides larger working volumes and minimises wicking through a special rounded square well design. Polypropylene is ideal for storage plates because it has a lower binding capacity, allowing for complete sample recovery.
Log in to see your contract pricing and availability.
  Remember me on this device
ORDER
Expand All / Collapse All
Description Colour Recommended working volume Pk VWR Catalog Number Quantity
DeepWell™ plate, 384 well, non treated, without lid Natural 5 - 240 µl 60 NUNC269390
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Select Product:
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty