Zirconyl nitrate 0,1% in hydrochloric acid 22% Reag. Ph. Eur. 1097201
Supplier: VWR Chemicals
|
Warning
|
Synonyms:
Zirconium dinitrate oxide
87966.180EA
14240
INR
87966.180
Zirconyl nitrate 0,1% in hydrochloric acid 22% Reag. Ph. Eur. 1097201
Zirconyl nitrate
Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.
Formula:
N₂O₇Zr MW: 231,23 g/mol Storage Temperature: Ambient |
CAS Number:
13826-66-9 EINECS: 237-529-3 UN: 1789 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...