Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phosphotungstic acid 22% in orthophosphoric acid 11% Reag. Ph. Eur. 1065200

Supplier: VWR Chemicals
Danger

Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.


Formula: H₃O₄₀PW₁₂
MW: 2880,05 g/mol
CAS Number: 1343-93-7
EINECS: 215-682-7
UN: 3264
ADR: 8,III

 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Plastic bottle 100 ml 87896.180
Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty