Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Avantor is taking action. Our response to the Novel Coronavirus outbreak.

Sulphanilic acid 0.02 mol/l in acetic acid (25%) Reag. Ph. Eur. 1086203

Supplier: VWR Chemicals
Danger
Synonyms: 4-Aminobenzenesulphonic acidAniline-4-sulphonic acid

87945.180EA 15647 INR InStock
87945.180
Sulphanilic acid 0.02 mol/l in acetic acid (25%) Reag. Ph. Eur. 1086203
Sulphanilic acid
Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.


Formula: 4-(H₂N)C₆H₄SO₃H
MW: 173,19 g/mol
Boiling Pt: 363 °C (1013 hPa)
Melting Pt: 279 °C
Density: 1,485 g/cm³ (25 °C)
Storage Temperature: ambient
MDL Number: MFCD00007886
CAS Number: 121-57-3
EINECS: 204-482-5
UN: 2790
ADR: 8,III
Merck Index: 13,09012
https://in.vwr.com/stibo/web/std.lang.all/76/05/2297605.jpg
Order Now

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty