Crystallisation microplates, Corning® CrystalEX™
Supplier: Corning
737-0158EA 0 INR
CORN3775 CORN3785 CORN3773
Crystallisation microplates, Corning® CrystalEX™
CrystalEX™ 96-well crystallisation microplates are optimised for high throughput protein crystal growth and screening. Reservoir and protein wells are positioned to be compatible with multi-head dispensing equipment.
96-well microplates: Features 96 large reservoir (reagent) wells and 96 corresponding protein wells, conical bottom protein wells allow improved centering of the protein drop. compatible with manual pipettors and automation, novel merged well design provides efficient vapor space for protein crystallisation.
384-well microplate: Meets ANSI/SBS standard footprint dimensions for microplates, ideal for fully automated crystal screening, features 192 reservoir wells and 192 corresponding protein wells, flat bottom protein wells are optimised for imaging of crystals.