KleenGuard™ MXRV safety goggles provide workers with reliable splash protection in an unventilated design.
- Universal size (one size fits most)
- Clear lenses allow maximum visible light transmission; ideal for indoor use
- Unventilated goggles provide protection from splashes and feature a curved lens for enhanced field of vision
- Polycarbonate lenses provide 99,9% UVA/UVB/UVC protection
- Anti-fog coating to prevent fogging; ideal for humid areas and temperature variances
KleenGuard™ MXRV safety goggles have a simple, sleek design and a curved lens for an enhanced vision field, while being roomy enough to fit over most prescription eye glasses. The lenses are clear, making them ideal for indoor use and maximum light transmission. They also feature anti-fog coating for humid environments and temperature variances.
Certifications: Meet the ANSI Z87.1+ standards for impact, with a D3 rating for droplet and splash protection.