Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pyronin Y, ultra pure grade

Supplier: VWR Chemicals
Synonyms: 3,6-Bis(dimethylamino)xanthylium chloridePyronin YPyronine Y

Used in Unna-Pappenheim stain to demonstrate RNA (red) in contrast to DNA (green). Also used for bacteria staining.

  • Colour index: 45005


Formula: C₁₇H₁₉ClN₂O
MW: 302,8 g/mol
Melting Pt: 250…260 °C
Storage Temperature: Ambient
MDL Number: MFCD00011725
CAS Number: 92-32-0
EINECS: 202-147-8
UN: 0000
Merck Index: 12,08190
Specification Test Results
Lambda max (0.1 g/500 ml then 1:100, Water) 543 – 553 nm

 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Amber bottle 10 g 0207-10G
Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty