Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phenylmethyl sulfonyl fluoride (PMSF), high purity grade

Supplier: VWR Chemicals
Danger
Synonyms: PMSFPhenylmethanesulphonyl fluoridePhenylmethylsulphonyl fluorideα-Toluenesulphonyl fluoride

An irreversible serine protease inhibitor of trypsin, chymotrypsin, kallikrein, subtilisin, and thrombin. Also inhibits the cysteine protease papain.


Formula: C₇H₇FO₂S
MW: 174,2 g/mol
Boiling Pt: 80 – 85 °C (0,3 torr)
Melting Pt: 91-92 °C
Storage Temperature: Ambient
CAS Number: 329-98-6
EINECS: 206-350-2
UN: 2923
ADR: 8 (6.1),III
Specification Test Results
Solubility (5%, EtOH) pass
Melting Point 91 – 95 °C
Thin Layer Chromatography (TLC) ONE SPOT
Solubility (5 %, EtOH) PASS
TLC ONE SPOT

Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
HDPE Bottle 25 g 0754-25G
Glass bottle 5 g 0754-5G
Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty