Would you like to log in again?
Chromium standard solution, 10,000 mg/l Cr in 10% hydrochloric acid (from (NH4)2Cr2O7) ARISTAR® standard for ICP
Supplier: VWR Chemicals
(NH₄)₂Cr₂O₇ in HCl 2-10%
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
Supplied with certificate of analysis.
ORDER
Expand All / Collapse All
Description Pack type Pk VWR Catalog Number Quantity
Chromium standard solution, 10,000 mg/l Cr in 10% hydrochloric acid (from (NH4)2Cr2O7) ARISTAR® standard for ICP Plastic bottle 500 ml 455254U
Chromium standard solution, 10,000 mg/l Cr in 10% hydrochloric acid (from (NH4)2Cr2O7) ARISTAR® standard for ICP Plastic bottle 100 ml 455252S
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty