Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium aluminate, coarse powder TECHNICAL

Supplier: VWR Chemicals
Danger



Formula: NaAlO₂
MW: 81,97 g/mol
Melting Pt: 1800 °C
Density: 1,5 g/cm³ (20 °C)
MDL Number: MFCD00801010
CAS Number: 1302-42-7
EINECS: 215-100-1
UN: 2812
ADR: 8,
Merck Index: 12,08715
Specification Test Results
Assay (calculated as Al2O3) 50 - 56 %
Identification Passes test

 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Plastic bottle for solids 1 kg 27663.290
Get a discount if you order 6 or more. Login in now for details.
Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty