Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Celite® Hyflo Super-cel®, filter aid, TECHNICAL

Supplier: VWR Chemicals
Warning

Diatomaceous silica for filtration.


MW: 60,08 g/mol
Boiling Pt: 2200 °C (1013 hPa)
Melting Pt: 1710 °C
Density: 2,3 g/cm³ (20 °C)
MDL Number: MFCD00132803
CAS Number: 61790-53-2
UN: 0000
Specification Test Results
Identification Passes test

Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Bucket (Plastic) 1 kg 24718.290
Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty