Would you like to log in again?
DECODRIDECON
Supplier: DECON LABORAT.
Surface active cleaning agent, in granular form, for use in laboratory, medical and industrial automatic glassware washing machines
Supplied as a finely milled powder, for use in automatic machines. PROPERTIES: Biodegradable, non-flammable, non-foaming and totally rinsable
Delivery information: This product is not available in all countries. Please check with your local VWR International office or supplier.
ORDER
Expand All / Collapse All
Description Pk Weight Pack type VWR Catalog Number Quantity
dri-decon® surface cleaning agent 5 kg 5 kg Plastic bottle 141-0001 Unavailable
unavailable This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty