Colchicine ≥98%
Supplier: VWR Chemicals
|
Danger
|
278052WEA
16647
INR
278052W
27805FM
Colchicine ≥98%
Colchicine
Formula:
C₂₂H₂₅NO₆ MW: 399,44 g/mol Melting Pt: 154…156 °C Density: 1,32 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00078484 CAS Number: 64-86-8 EINECS: 200-598-5 UN: 1544 ADR: 6.1,I Merck Index: 12,02536 |
Specification Test Results
Assay | Min. 98% (HPLC) |
Identity (IR-spectrum) | Passes test |
Specific rotation (a 20/D; 1%; water) | -250 to -220°C |
Water (according to Karl Fischer) | Max 2% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...