Phenolphthalein 0,5% in ethanol 50% for milk analysis
|
Danger
|
Synonyms:
3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one
M-Clarity™ quality level = MQ100
210296BDP
Phenolphthalein 0,5% in ethanol 50% for milk analysis
Phenolphthalein
Formula:
C₂₀H₁₄O₄ MW: 318,33 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00005913 CAS Number: 77-09-8 EINECS: 201-004-7 UN: 1170 ADR: 3,III |
Specification Test Results
pH (20°C) | 6.5 - 7.0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...