Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sulphurous acid 4,5 - 6% in aqueous solution

Supplier: VWR Chemicals
Danger
Synonyms: Sulphur dioxide solution



Formula: H₂O₃S
MW: 82,08 g/mol
Density: 1,03 g/cm³ (20 °C)
MDL Number: MFCD00011335
CAS Number: 7782-99-2
EINECS: 231-973-1
UN: 1833
ADR: 8,II
Specification Test Results
Assay 5.0 - 6.0 %
Cl (Chloride) Max. 0.0005 %
Ag (Silver) Max. 0.000002 %
Al (Aluminium) Max. 0.000005 %
As (Arsenic) Max. 0.00005 %
Ba (Barium) Max. 0.000005 %
Be (Beryllium) Max. 0.000002 %
Bi (Bismuth) Max. 0.000010 %
Ca (Calcium) Max. 0.0001 %
Cd (Cadmium) Max. 0.000005 %
Co (Cobalt) Max. 0.000002 %
Cr (Chromium) Max. 0.000002 %
Cu (Copper) Max. 0.000002 %
Fe (Iron) Max. 0.00005 %
Ge (Germanium) Max. 0.000005 %
K (Potassium) Max. 0.000010 %
Li (Lithium) Max. 0.000002 %
Mg (Magnesium) Max. 0.000010 %
Mn (Manganese) Max. 0.000002 %
Mo (Molybdenum) Max. 0.000002 %
Na (Sodium) Max. 0.00005 %
Ni (Nickel) Max. 0.000002 %
Pb (Lead) Max. 0.000002 %
Sr (Strontium) Max. 0.000002 %
Ti (Titanium) Max. 0.000010 %
Tl (Thallium) Max. 0.000005 %
V (Vanadium) Max. 0.000005 %
Zn (Zinc) Max. 0.000005 %
Zr (Zirconium) Max. 0.000010 %
Residue on ignition Max. 0.002 %

 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle 2,5 l 191655T Unavailable
unavailable This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty