You Searched For: Wipes for Delicate Tasks


9  results were found

SearchResultCount:"9"

Sort Results

List View Easy View

Rate These Search Results

Supplier: VWR Collection
Description: Absorbent paper, suitable for cleaning and drying glass containers, tubes, microscopic plates, bottles, and more.

Catalog Number: (KIMB21905-026_U)
Supplier: KIMBERLY CLARK
Description: KIMWIPES* are suitable for general multipurpose and delicate task cleaning. The creped surface picks up and holds oil, dust, and liquids from glass, p...
UOM: 1 * 280 items


Supplier: KIMBERLY CLARK
Description: These single ply, light-duty wipes can handle a variety of delicate tasks and easily wipe up liquid and dust.

Supplier: KIMBERLY CLARK
Description: These high quality wipes, made from 100% pure fibre, are suitable for a variety of precision wiping tasks such as the cleaning of delicate glassware, ...

Catalog Number: (115-2063)
Supplier: KIMBERLY CLARK
Description: Durable uniformly reinforced towels for demanding jobs. Can be used in medium to heavy duty applications.
UOM: 1 * 912 items


Catalog Number: (KIMB34705)
Supplier: KIMBERLY CLARK
Description: This 2-ply wipe offers a unique combination of low-lint tissue softness and multi-ply absorbency. Perfect for a multitude of delicate tasks.
UOM: 1 * 15 items


Catalog Number: (KIMB33560)
Supplier: KIMBERLY CLARK
Description: These PP wipes are engineered for critical applications involving solvents, inks, and paints. Typical applications include: furniture finishing, engin...
UOM: 1 * 66 items


Catalog Number: (KIMB34120_U)
Supplier: KIMBERLY CLARK
Description: These light-duty wipes can handle a variety of delicate tasks and easily wipe up liquid and dust.
UOM: 1 * 8.400 items


Catalog Number: (VWRU21910-105)
Supplier: VWR Collection
Description: These bleached cloths made from 100% pure cotton of reagent-grade quality and purity are ideal for wiping, cleaning, or drying cuvettes, delicate glas...
UOM: 1 * 60 items


Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 9 of 9
no targeter for Bottom