Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

sterile-cleanroom-wipes

SearchPresentationType-VERTICAL
 
 
SearchResultCount:"11"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  VWR Collection
Description:   These cleanroom wipes are made of polyester and cellulose.
UOM:  1 * 100 items
Supplier:  VWR Collection
Description:   These wipes made from double woven polyester fabric with close-cut edges are washed in ISO Class 4 cleanrooms to minimise contamination by particles a...
Supplier:  Texwipe
Description:   These single-ply, heavyweight polyester sealed-border wipes offer the same capacity sorption as a two-ply wipe. They are suitable for all general purp...
Supplier:  Texwipe
Description:   SterileWipe™ LP is a 100% continuous-filament cut edge polyester wipe that is low in particle generation and extractable levels. It provides excellen...
UOM:  1 * 500 items
Supplier:  Texwipe
Description:   Vertex® wipes are the cleanest, most consistent and most rigorously quality tested cleanroom wipes.
Supplier:  Texwipe
Description:   SterileWipe™ HS II is a non woven cellulose/polyester-blend wipe offering both durability and absorbency. It is an excellent general purpose sterile w...
UOM:  1 * 500 items
Supplier:  Texwipe
Description:   SterileWipe™ AS 10 is a 2 ply, double knit construction, 100% continuous filament polyester wipe that offers high sorbency in a sealed-border wipe to ...
UOM:  1 * 100 items
Supplier:  Texwipe
Description:   SterileWipe™ LP 10 is a 100% continuous-filament polyester wipe with thermally sealed borders that have been designed to control particle and fiber co...
UOM:  1 * 500 items
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 8  of 11
  1  2  Next