Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • In Stock (2)
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

You Searched for: "standard-incubators"

https://in-bot.vwr.com/stibo/smallweb/std.lang.all/98/45/18989845.jpg
SearchPresentationType-VERTICAL
 
 
SearchResultCount:"7"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  VWR Collection
Description:   These incubators with natural air convection feature a stainless steel inner chamber, a powder coated sheet exterior and an internal glass door. All m...
Supplier:  Binder
Description:   Two versions of these incubators with electronically controlled APT.line™ preheating chamber are available depending on the application: The BD series...
Supplier:  THERMO BRAND
Description:   Heratherm® General Protocol incubators feature a smooth stainless steel inner chamber with an internal glass door, allowing samples to be viewed witho...
Supplier:  Binder
Description:   Avantgarde.Line incubators ensure stable and reliable incubation of microbiological cultures and guarantee reproducible results for routine tests in t...
Supplier:  MEMMERT
Description:   The Generation 2012 range has natural and forced convection models with a temperature range up to +80 °C. There is a choice of two different displays,...
Catalog Number: (390-0854)
Supplier:  GENLAB
Description:   Reliable, efficient, general purpose incubators, to suit most biological analysis, research and general laboratory applications. These units from Genl...
UOM:  1 * 1 items
Supplier:  Panasonic Biomedical
Description:   Heated incubators with stainless steel interior and scratch resistant baked-on acrylic finish incorporate microprocessor controller with PID heater co...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 7  of 7