Pipette tips, superstack, non sterile
VWRI613-6466EA 0 INR
VWRI613-6466
Pipette tips, superstack, non sterile
Thin wall crown design provides optimum seal and universal fit on all standard designed single and multichannel pipettors.
  • More flexible than traditional, rigid pipette tip - better ergonomic design
  • Manufactured using both recycled and recyclable materials
  • Ink jet identification used to preserve recyclability
  • All trays are clearly labelled with volume and part number
  • Dense polyethylene barrier protects against aerosol and liquid contamination and provides the highest levels of purity
  • Fully recyclable trays and racks
Graduation marks on all tips for quick volume reference and certified-free of detectable DNase, RNase, and human DNA.