CyMet ERMA III DIFF for hematology, J.T.Baker®
Supplier: Avantor
3416-00EA
0
INR
3416-00
3416.05
CyMet ERMA III DIFF for hematology, J.T.Baker®
CyMet
Lysing reagent for hematology
Caution: In vitro diagnostic medical devices only for professional use.
Specification Test Results
Conductivity (mS) | 3.10 - 4.10 |
Osmolality (mOsm/kg) | 35 - 45 |
Performance Test * | passes test |
pH at 25°C | 10.0 - 10.4 |
*) All parameters except mid cells: Test/Ref ratio > 95% and <105%. Correlation coefficient: > 0.95 | |
Test results are valid at time of lot release. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...