CyMet Micro for hematology CN-free, J.T.Baker®
Supplier: Avantor
3863-00EA
0
INR
3863-00
CyMet Micro for hematology CN-free, J.T.Baker®
CyMet
Lysing reagent for hematology
Caution: In vitro diagnostic medical devices only for professional use.
Specification Test Results
Conductivity (mS) | 3.58 - 4.58 |
Osmolality (mOsm/kg) | 51 - 61 |
Performance Test | passes test |
pH at 25°C | 5.50 - 6.80 |