Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 1:00 am CET Saturday through 0:30 am Monday. Orders placed will be processed after maintenance is complete.

Ninhydrin reagent (including 1 litre buffer)

Supplier: VWR Chemicals
Warning
Synonyms: 2,2-Dihydroxyindan-1,3-dionTrioxohydrindene monohydrateNinhydrin monohydrate2,2-Dihydroxy-1,3-indanedioneIndanetrione hydrate1,2,3-Indantrione monohydrate

HPLCN89002.EA 82746 INR InStock
HPLCN89002.
Ninhydrin reagent (including 1 litre buffer)
Ninhydrin


Formula: C₉H₆O₄
MW: 178,14 g/mol
Boiling Pt: 351 °C (1013 hPa)
Melting Pt: 250 to 258 °C
Density: 0,862 g/cm³ (20 °C)
Storage Temperature: ambient
MDL Number: MFCD00003791
CAS Number: 485-47-2
EINECS: 207-618-1
UN: 0000
Merck Index: 13,06582
https://in.vwr.com/stibo/web/std.lang.all/70/01/2297001.jpg
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty