Octyl phenol ethoxylate ≥90.0%, Technical Grade, J.T.Baker®
|
Warning
|
Synonyms:
alkylaryl polyether alcohol
, Polyethylene glycol tert-octylphenyl ether
Management of Change (MOC) category = N
X199-05EA
0
INR
X199-05
X199-03
X199-01
Octyl phenol ethoxylate ≥90.0%, Technical Grade, J.T.Baker®
Octyl phenol ethoxylate
Caution: Keep container tightly closed. Minimum recommended storage temperature is 23 °C.
Note: A precipitate may form on prolonged storage. Heat to 40 to 50 °C with agitation to render homogeneous solution.
Density:
1,07 g/cm³ Flash Pt: 251,00 °C |
CAS Number:
9036-19-5 |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
EP - Acidity or Alkalinity | Passes Test |
EP - Cloud Point | 63 - 70 °C |
EP - Hydroxyl value, mg KOH/g | 85 - 101 |
EP - Identification A (IR) | Passes Test |
EP - Identification B (Cloud Point) | Passes Test |
EP - Limit of Dioxane | ≤10 ppm |
EP - Limit of Free Ethylene Oxide | ≤1 ppm |
EP - Total Ash | ≤0.4 % |
EP - Water | ≤1 % |
NF - Acid Value | ≤0.2 |
NF - Assay (LC) | 90.0 - 110.0 % |
NF - Content of Free PEGs | ≤1 % |
NF - Heavy Metals (as Pb) | ≤20 ppm |
NF - Hydroxyl value, mg KOH/g | 85 - 101 |
NF - Identification A (IR) | Passes Test |
NF - Identification B (LC) | Passes Test |
NF - Limit of Dioxane | ≤10 ppm |
NF - Limit of Free Ethylene Oxide | ≤5 ppm |
NF - Peroxide value, meq/kg | ≤10 |
NF - Residue on ignition | |
NF - Water | ≤0.5 % |
Acetaldehyde | ≤5 ppm |
Bacterial endotoxins, IU/mL | ≤100 |
Chloride (Cl), For Information Only | |
Ethylene Glycol | ≤600 ppm |
Formaldehyde | ≤5 ppm |
Limit of ethyl acetate | ≤1000 ppm |
Total Aerobic Microbial Count | ≤1000 cfu/g |
Total Coliform | Negative |
Arsenic (As), For Information Only | |
Cadmium (Cd), For Information Only | |
Lead (Pb), For Information Only | |
Mercury (Hg), For Information Only | |
Aluminum (Al), For Information Only | |
Barium (Ba), For Information Only | |
Boron (B), For Information Only | |
Chromium (Cr), For Information Only | |
Copper (Cu), For Information Only | |
Iron (Fe), For Information Only | |
Magnesium (Mg), For Information Only | |
Manganese (Mn), For Information Only | |
Nickel (Ni), For Information Only | |
Strontium (Sr), For Information Only | |
Tungsten (W), For Information Only | |
Zinc (Zn), For Information Only | |
Calcium (Ca), For Information Only | |
Potassium (K), For Information Only | |
Sodium (Na), For Information Only | |
Residual Solvents: Only the Class 2 solvents ethylene glycol and | |
dioxane are expected to be present as by-products of the manufacturing | |
process. The Class 3 solvent ethyl acetate is used in the production | |
process. Content of these 3 solvents are batch tested and reported as | |
impurities. No other Class 1, 2, 3 or any other residual solvent is | |
expected to be present. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...