Benzyl alcohol ≥99.8% for HPLC
Supplier: RANKEM (A AVANTOR BRAND)
|
Warning
|
Synonyms:
Phenylmethanol
, Phenylcarbinol
RANKB0211EA
4719
INR
RANKB0211
RANKB0212
Benzyl alcohol ≥99.8% for HPLC
Benzyl alcohol
Formula:
C₆H₅CH₂OH MW: 108,14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00004599 CAS Number: 100-51-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...