Calcium sulphate, Drierite™ (8 Mesh), Macron Fine Chemicals™
Supplier: Avantor Performance Materials
|
Danger
|
MACR3378-04EA
0
INR
MACR3378-04
Calcium sulphate, Drierite™ (8 Mesh), Macron Fine Chemicals™
Calcium sulphate
Formula:
CaO₄S MW: 136,14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00010912 CAS Number: 7778-18-9 EINECS: 231-900-3 UN: 3077 ADR: 9,III |
Specification Test Results
For Laboratory,Research,or Manufacturing Use |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...