Perchloric acid 70% AR, Rankem®
Supplier: RANKEM (A AVANTOR BRAND)
|
Danger
|
RANKP0068EA
0
INR
RANKP0068
RANKP0067
RANKP0065
Perchloric acid 70% AR, Rankem®
Perchloric acid
Formula:
HClO₄ MW: 100,46 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011325 CAS Number: 7601-90-3 UN: 1873 ADR: 5.1,I |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...