BeneSphera™, ELISA Kit, HIV-1/2
Supplier: Avantor
BeneSphera™
BENP15000992EA 2550 INR
BENP15000992
BeneSphera™, ELISA Kit, HIV-1/2
Assays ELISAs
BeneSphera™ HIV 1/2 is an in vitro enzyme immunoassay for the qualitative determination of antibodies to HIV-1 and HIV-2 in Human Serum or Plasma.
To evaluate the specificity of the BeneSphera™ HIV 1/2 a total of 1500 samples were tested for Anti-HIV-1/Anti HIV-2, of 1500 samples, 8 were found to be positive with BeneSpheraTM HIV 1/2.
But these sera were all negative in HIV Blot kit. The specificity of the BeneSphera™ HIV 1/2 is 99,5%, The newly developed kit consists of recombinant HIV antigens and conjugates, i.e. recombinant HIV1 (gp-41, p24) and HIV-2 antigen (gp36).
To test reproducibility of BeneSphera™ HIV 1/2, 3 plates from 3 production batches each were randomly selected and tested against a Q.C. panel of HIV-1, HIV-2 positive and negative samples.
Delivery information: The kit contains HIV 1/2 Antigen coated microstrips, Plate sealer in zip seal polybag, Sample diluent, Negative control, Positive control, Conjugate concentrate (101×), Conjugate diluent, TMB chromogen in DMSO, Substrate buffer, Washing solution concentrate (20×), Stop solution, Empty zip seal polybag.