Rankem®, Membrane Filters, Nylon
Supplier: Avantor
RANKNY0247EA 2500 INR
RANKNY0247 RANKNY0225 RANKNY45142 RANKNY4513 RANKNY4547 RANKNY4525 RANKNY0213 RANKNY02293
Rankem®, Membrane Filters, Nylon
Filters Membrane Filters
These nylon membranes are used for filtration of alkaline solutions and organic solvents. Designed for particle removing filtration of water, aqueous solutions and solvents for analytical determination and filtration of mobile phase solvents for HPLC analysis.
  • Hydrophillic
  • Chemically resistant to alkaline solutions and organic solvents
Used in pressure filtration devices, filtration of biological, pharmaceutical and environmental solutions, particle removal from solvents, filtration of air, gases or chemicals microbiological quality control.