Haematoxylin solution (Harris), Gurr™ for microscopy mercury-free
Synonyms:
Hydroxybrasilin
, DcH-48
, Natural Black I
351945SEA
11130
INR
351945S
Haematoxylin solution (Harris), Gurr™ for microscopy mercury-free
Haematoxylin staining solutions
Formula:
C₁₆H₁₄O₆ MW: 302,28 g/mol |
MDL Number:
MFCD00078111 CAS Number: 517-28-2 |
Specification Test Results
Cyanophilic cytoplasm | Blue-green |
Eosinophilic cytoplasm | Pink |
Nuclei | Blue to dark violet |
Suitable for microscopy (vaginal smear) | Passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...