Chemiluminescence analyser, Maglumi 800
Supplier: Mindray
Maglumi
SNIB23020003EA 1300000 INR
SNIB23020003
Chemiluminescence analyser, Maglumi 800
Clinical Diagnostic Systems Clinical Analysers
Maglumi 800 is a random access CLIA analyser.
  • Throughput: Up to 180 tests/hour
  • Reagents onboard: 9
  • Sample loading: Up to 40 samples
  • Integrated reagent kit ready to use
  • Long term calibration stability
Delivery information: Includes all necessary calibrators and controls.