BeneSphera™, Malaria Plasmodium vivax/Plasmodium falciparum Rapid Test
Supplier: Avantor
BeneSphera™
BENP15000907EA 1500 INR
BENP15000907 BENP15000906 BENP15001258 BENP15001260
BeneSphera™, Malaria Plasmodium vivax/Plasmodium falciparum Rapid Test
Clinical Diagnostic Supplies Clinical Reagents and Kits
BeneSphera™ Malaria Plasmodium vivax/Plasmodium falciparum Rapid Test is for the differential diagnosis of Plasmodium vivax and Plasmodium falciparum.
  • Detection of pLDH of Plasmodium vivax and HRP-II of Plasmodium falciparum
  • Whole blood specimen
  • Takes 20 to 30 minutes for test result