ELISA microplate washer, BeneSphera™ W-21
Supplier: BeneSphera
BENP20001754EA 150300 INR
BENP20001754
ELISA microplate washer, BeneSphera™ W-21
Microplate Washers
The BeneSphera™ W-21 ELISA washer is suitable for flat, U-bottom or V-bottom plates and strip washing.
  • Large memory to store up to 50 user programmable wash protocols
  • Liquid level warning
  • Includes an 8-way manifold
  • Emergency stop available
  • Low residual volume
  • Large LCD display with Win CE compatible operating system