Flow cytometer and cell count analyser, Moxi Flow™
Supplier: Orflo
ORFLMXF002EA 0 INR
ORFLMXF002
Flow cytometer and cell count analyser, Moxi Flow™
Cell Counters Flow Cytometers
Moxi Flow™ is a fully automated, cassette-based flow cytometer that combines unparalleled ease of use with the precision and accuracy normally only associated with more expensive flow cytometers. This ultra-small instrument uses patented microfluidic thin-film cassettes that enable automatic load and measure operation. The Moxi Flow is also the only flow cytometer that auto-aligns the laser to each cassette to enable highly repeatable and robust results every time.
  • Cell count with >95% accuracy
  • Viability with PI
  • Apoptosis with Annexin V
  • Precise cell/particle (Coulter Principle)
  • Blood analysis (RBC counts, WBC counts and CD4 counts)
  • Somatic cell counts in milk
Open flow cytometry assay miniature open platform can be used for common cell assay needs, or to develop sophisticated, custom assays right at your fingertips.
Delivery information: Includes USB power cord, US style USB power adapter, and type MF-S cassette pack.