Calcium fluoride ≥95.0% (by EDTA titration), powder, BAKER ANALYZED®, J.T.Baker®

Synonyms: FluoriteFluorspar

1354-07EA 0 INR
1354-07
Calcium fluoride ≥95.0% (by EDTA titration), powder, BAKER ANALYZED®, J.T.Baker®
Calcium fluoride
Formula: CaF₂
MW: 78,07 g/mol
Density: 3,18 g/cm³
MDL Number: MFCD00010907
CAS Number: 7789-75-5

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (CaF₂) (by EDTA titrn) ≥95.0 %
Chloride (Cl) ≤0.1 %
Sulfate (SO₄) ≤0.05 %
Heavy Metals (as Pb) ≤0.005 %
Iron (Fe) ≤0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR