Agarose PFGE, Ultrapure Bioreagent, J.T.Baker®
4063-05EA
31814
INR
4063-05
Agarose PFGE, Ultrapure Bioreagent, J.T.Baker®
Agarose
For rapid resolution of DNA greater than 1000 bp
Density:
0,90 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00081294 CAS Number: 9012-36-6 |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Loss on Drying at 70°C (in vacuo) | ≤7.0 % |
Sulfate (SO₄) | ≤0.12 % |
Electroendosmosis (EEO)(-m^r) | ≤0.12 % |
Gel Strength, g/cm² (MCl, 1.0%) | ≥2000 |
Gelling Temperature of 1.5% w/w Solution | 34.5 - 37.5 °C |
Melting Temperature of 1.5% w/w Solution | 86.5 - 89.5 °C |
DNase Activity (None Detected) | Passes Test |
RNase Activity | Passes Test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...