Calmagite, J.T.Baker®

E280-03EA 51392 INR
E280-03
Calmagite, J.T.Baker®
Calmagite
Formula: C₁₇H₁₄N₂O₅S
MW: 358,37 g/mol
MDL Number: MFCD00011656
CAS Number: 3147-14-6

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Sensitivity as Metal Indicator Passes Test
Identification (by IR) Passes Test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR