Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Avantor is taking action. Our response to the Novel Coronavirus outbreak.

Manganese, plasma standards, BAKER INSTRA-ANALYZED® reagent, J.T.Baker®

Supplier: Avantor Performance Materials
BAKER INSTRA-ANALYZED®
5793-04EA 0 INR InStock
5793-04
Manganese, plasma standards, BAKER INSTRA-ANALYZED® reagent, J.T.Baker®
Standards Cationic Standards Manganese Standards
Plasma standards.

  • Prepared from the highest purity raw material 99,999%
  • Assay value accurate within ±0,3% of the assay value
  • Certificate of analysis lists the trace impurity level in µg/ml for over 65 elements

Caution: 10,000 µg/ml= (1.00% w/v)= 10000 mg/l=10000 ppm

Order Now
 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty