Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Avantor is taking action. Our response to the Novel Coronavirus outbreak.

Iodine monochloride solution 0.215 - 0.225 N in acetic acid, BAKER ANALYZED® PharmaTest™ USP Test Solution (TS), J.T. Baker®

Supplier: Avantor Performance Materials
Danger
Synonyms: Iodochloride

5924-02EA 0 INR InStock
5924-02
Iodine monochloride solution 0.215 - 0.225 N in acetic acid, BAKER ANALYZED® PharmaTest™ USP Test Solution (TS), J.T. Baker®
Iodine monochloride


Formula: ClI
MW: 162,36 g/mol
Boiling Pt: 97.3 °C (1013 hPa)
Melting Pt: 26 °C
Density: 3,1 g/cm³ (29 °C)
Flash Pt: 40 °C
Storage Temperature: ambient
MDL Number: MFCD00011354
CAS Number: 7790-99-0
EINECS: 232-236-7
UN: 2920
ADR: 8, 3,II
Merck Index: 13,05039
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Normality 0.215 - 0.225
Meets test solution (T.S.) specifications for testing USP/NF
monographs.
Product Information (not specifications):
Appearance (Deep, orange-brown solution)

 

ORDER

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty