Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Avantor is taking action. Our response to the Novel Coronavirus outbreak.

tert-Butyl methyl ether ≥99.0% (by GC, corrected for water content), BAKER ANALYZED® ACS, J.T. Baker®

Supplier: Avantor Performance Materials
Danger

9034-03EA 0 INR InStock
9034-03
tert-Butyl methyl ether ≥99.0% (by GC, corrected for water content), BAKER ANALYZED® ACS, J.T. Baker®
tert-Butyl methyl ether


Formula: (CH₃)₃COCH₃
MW: 88,15 g/mol
Boiling Pt: 55.2 °C (1013 hPa)
Melting Pt: –108,6 °C
Density: 0,74 g/cm³
Flash Pt: –27,00 °C
Storage Temperature: ambient
MDL Number: MFCD00008812
CAS Number: 1634-04-4
EINECS: 216-653-1
UN: 2398
ADR: 3,II
Merck Index: 13,06059
https://in.vwr.com/stibo/web/std.lang.all/77/75/2297775.jpg
Order Now
Specification Test Results
Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (C₅H₁₂O) (by GC, corrected for water) ≥ 99.0 %
ACS - Color (APHA) ≤ 10
ACS - Peroxide (as H₂O₂) ≤ 1 ppm
ACS - Residue after Evaporation ≤ 0.001 %
ACS - Water (H₂O) ≤ 0.05 %

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty